Hexacarbonyltungsten, 97%, Alfa Aesar™

Manufacturer: Alfa Aesar 4320509

Quantity 10g

 View more versions of this product

Catalog No. AA4320509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14040-11-0
    Physical Form Crystalline
    Assay Percent Range 97%
    Molecular Formula C6O6W
    Formula Weight 351.91
    MDL Number MFCD00011462
    Synonym Tungsten hexacarbonyl

    Specifications

    Formula Weight 351.91
    Quantity 10g
    Color White
    Physical Form Crystalline
    Assay Percent Range 97%
    Odor Nearly odorless
    Molecular Formula C6O6W
    MDL Number MFCD00011462
    CAS 14040-11-0
    Synonym Tungsten hexacarbonyl
    Vapor Pressure 1.596hPa (1mm Hg) at 67°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit