Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexacarbonyltungsten, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar 4320518

Quantity 50g

 View more versions of this product

Catalog No. AA4320518


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

14040-11-0
97%
351.91
Tungsten hexacarbonyl
Crystalline
C6O6W
MFCD00011462

Specifications

50g
Crystalline
Nearly odorless
351.91
Tungsten hexacarbonyl
White
14040-11-0
97%
C6O6W
MFCD00011462
1.596hPa (1mm Hg) at 67°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit