Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexachloroethane, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
67-72-1
C2Cl6
236.74
VHHHONWQHHHLTI-UHFFFAOYSA-N
hexachloroethane,perchloroethane,ethane, hexachloro,avlothane,carbon hexachloride,hexachlorethane,mottenhexe,distokal,distopan,distopin
6214
CHEBI:39227
1,1,1,2,2,2-hexachloroethane
C(C(Cl)(Cl)Cl)(Cl)(Cl)Cl
67-72-1
C2Cl6
236.74
VHHHONWQHHHLTI-UHFFFAOYSA-N
hexachloroethane,perchloroethane,ethane, hexachloro,avlothane,carbon hexachloride,hexachlorethane,mottenhexe,distokal,distopan,distopin
6214
CHEBI:39227
1,1,1,2,2,2-hexachloroethane
C(C(Cl)(Cl)Cl)(Cl)(Cl)Cl
C-X bond formation reagent
Water 0.03% max.
Color White
Density 2.0910g/mL
Quantity 1kg
Infrared Spectrum Authentic
Specific Gravity 2.091
Melting Point 184.0°C to 190.0°C
Packaging Plastic bottle
Assay Percent Range 98.5% min. (GC)
Linear Formula Cl3CCCl3
Beilstein 01,87
Merck Index 15,4715
Solubility Information Solubility in water: 0.05g/L (22°C). Other solubilities: soluble in most common organic solvents,>50g/100mL ethanol
Formula Weight 236.74
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Chemical Name or Material Hexachloroethane
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN

GHS Signal Word: Warning

EINECSNumber : 200-666-4