Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexachloroethane, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
67-72-1
C2Cl6
236.722
VHHHONWQHHHLTI-UHFFFAOYSA-N
hexachloroethane,perchloroethane,ethane, hexachloro,avlothane,carbon hexachloride,hexachlorethane,mottenhexe,distokal,distopan,distopin
6214
CHEBI:39227
1,1,1,2,2,2-hexachloroethane
C(C(Cl)(Cl)Cl)(Cl)(Cl)Cl
67-72-1
C2Cl6
236.722
VHHHONWQHHHLTI-UHFFFAOYSA-N
hexachloroethane,perchloroethane,ethane, hexachloro,avlothane,carbon hexachloride,hexachlorethane,mottenhexe,distokal,distopan,distopin
6214
CHEBI:39227
1,1,1,2,2,2-hexachloroethane
C(C(Cl)(Cl)Cl)(Cl)(Cl)Cl
C-X bond formation reagent
Packaging Plastic bottle
Color White
Infrared Spectrum Authentic
Melting Point 184.0°C to 190.0°C
Density 2.0910g/mL
Quantity 2.5kg
Specific Gravity 2.091
Water 0.03% max.
Assay Percent Range 99%
Linear Formula Cl3CCCl3
Formula Weight 236.74
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Chemical Name or Material Hexachloroethane
Show More Show Less