Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexacosane, ≥98% (GC), MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 521831G
Description
Featured Industries: PetroleumSpecifications
630-01-3 | |
Analytical Standard | |
MFCD00009354 | |
≥98% (GC) | |
55°C to 58°C; 55°C to 58°C |
1 g | |
CH3(CH2)24 CH3 | |
366.71 | |
Analytical Standard | |
Neat |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction