Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexadecane, Reagents
Click to view available options
Quantity:
100 mL
450 mL
Specifications
Specifications
| Boiling Range | 287°C |
| Color | Colorless |
| Boiling Point | 287°C |
| Packaging | Glass Bottle |
| Quantity | 450 mL |
| Specific Gravity | 0.773 |
| Grade | Reagent |
| Physical Form | Liquid |
| Chemical Name or Material | Hexadecane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction