Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecanedioic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
2.5g
505-54-4
C16H30O4
286.412
QQHJDPROMQRDLA-UHFFFAOYSA-N
thapsic acid,1,14-tetradecanedicarboxylic acid,1,16-hexadecanedioic acid,hexadecane-1,16-dioic acid,1,14-tetradecane dicarboxylic acid,n-tetradecane-.omega.,.omega.'-dicarboxylic acid,dicarboxylic acid c16,1,16-hexadecanedioate,thapsate,thaspic acid
10459
CHEBI:73722
hexadecanedioic acid
C(CCCCCCCC(=O)O)CCCCCCC(=O)O
505-54-4
C16H30O4
286.412
QQHJDPROMQRDLA-UHFFFAOYSA-N
thapsic acid,1,14-tetradecanedicarboxylic acid,1,16-hexadecanedioic acid,hexadecane-1,16-dioic acid,1,14-tetradecane dicarboxylic acid,n-tetradecane-.omega.,.omega.'-dicarboxylic acid,dicarboxylic acid c16,1,16-hexadecanedioate,thapsate,thaspic acid
10459
CHEBI:73722
hexadecanedioic acid
C(CCCCCCCC(=O)O)CCCCCCC(=O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 1g
Assay Percent Range 97%
Linear Formula HO2C(CH2)14CO2H
Beilstein 02, 733
Formula Weight 286.4
Physical Form Fine Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material Hexadecanedioic acid, 97%
Show More Show Less

EINECSNumber : 208-013-5

TSCA : TSCA