Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyl Chloroformate 98.0+%, TCI America™
SDP

Catalog No. C105325ML
Click to view available options
Quantity:
25 mL

Chemical Identifiers

CAS 26272-90-2
Molecular Formula C17H33ClO2
Molecular Weight (g/mol) 304.899
MDL Number MFCD00035954
InChI Key HOQUWXSARQBQCW-UHFFFAOYSA-N
Synonym Cetyl Chloroformate, Chloroformic Acid Cetyl Ester, Chloroformic Acid Hexadecyl Ester
PubChem CID 97633
IUPAC Name hexadecyl carbonochloridate
SMILES CCCCCCCCCCCCCCCCOC(=O)Cl

Specifications

Color Yellow
Quantity 25 mL
UN Number 3277
Formula Weight 304.90
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Hexadecyl Chloroformate
EINECSNumber (2)-3549
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.