Learn More
Hexafluorosilicic acid, 23% w/w aq. soln., Thermo Scientific Chemicals
Description
It is mainly produced as a precursor to aluminum trifluoride and synthetic cryolite. It is commonly used as a source of fluoride for water fluoridation. Hexafluorosilicic acid is also converted to a variety of useful hexafluorosilicate salts. The potassium salt is used in the production of porcelains, the magnesium salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White through Yellow |
Physical Form | Liquid |
Quantity | 500 g |
UN Number | UN1778 |
Merck Index | 14,4182 |
Solubility Information | Fully miscible in water. |
Formula Weight | 144.09 |
Concentration or Composition (by Analyte or Components) | 23% w/w aq. soln. |
Density | 1.223g/mL |
Chemical Name or Material | Hexafluorosilicic acid |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FLUOROSILICIC ACID |
EINECSNumber | 241-034-8 |
RTECSNumber | VV8225000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.