Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethonium bromide hydrate, +98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
55-97-0
C12H30Br2N2
362.194
MFCD00149988
FAPSXSAPXXJTOU-UHFFFAOYSA-L
hexamethonium bromide,hexamethonium dibromide,simpatoblock,gangliostat,esametina,hexameton,vegolysen,vegolysin,bistrium bromide,hexonium dibromide
5938
trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide
C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-]
55-97-0
C12H30Br2N2
362.194
MFCD00149988
FAPSXSAPXXJTOU-UHFFFAOYSA-L
hexamethonium bromide,hexamethonium dibromide,simpatoblock,gangliostat,esametina,hexameton,vegolysen,vegolysin,bistrium bromide,hexonium dibromide
5938
trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide
C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-]
Color White
Quantity 25g
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 10% Max.
Assay Percent Range 98+%
Linear Formula [-(CH2)3N(CH3)3Br]2·H2O
Beilstein 04, IV, 1323
Merck Index 15, 4724
Formula Weight 362.2
Physical Form Crystalline Powder and Agglomerates
Percent Purity ≥98.0% (on dry substance)
Chemical Name or Material Hexamethonium bromide hydrate
Show More Show Less