Alfa Aesar™ Hexamethylacetone, 98%

Manufacturer: Alfa Aesar™ H5346406

Quantity 5g

 View more versions of this product

Catalog No. AAH5346406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 815-24-7
    Molecular Formula C9H18O
    Formula Weight 142.24
    MDL Number MFCD00008845
    UN Number UN1224
    Beilstein 1701122
    Synonym Di-tert-butyl ketone; 2,2,4,4-Tetramethyl-3-pentanone
    Chemical Name or Material Hexamethylacetone

    Specifications

    Formula Weight 142.24
    MDL Number MFCD00008845
    Synonym Di-tert-butyl ketone; 2,2,4,4-Tetramethyl-3-pentanone
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 34°C (93°F)
    Quantity 5g
    TSCA TSCA
    Molecular Formula C9H18O
    CAS 815-24-7
    UN Number UN1224
    Beilstein 1701122
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
    Boiling Point 152° to 153°C
    Chemical Name or Material Hexamethylacetone
    Assay 98%

    Description

    Applications
    Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit