Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hexamethylcyclotrisiloxane, 97%

Click to view available options
Quantity:
25g
100g
500g
541-05-9
C6H18O3Si3
222.462
MFCD00005943
HTDJPCNNEPUOOQ-UHFFFAOYSA-N
hexamethylcyclotrisiloxane,cyclotrisiloxane, hexamethyl,dimethylsiloxane cyclic trimer,unii-epv75l8o0r,ccris 1326,sdk 10,hexamethylcyclohexasiloxane,epv75l8o0r,cyclotrisiloxane, 2,2,4,4,6,6-hexamethyl,dsstox_cid_7185
10914
2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-trioxatrisilinane
C[Si]1(O[Si](O[Si](O1)(C)C)(C)C)C
541-05-9
C6H18O3Si3
222.462
MFCD00005943
HTDJPCNNEPUOOQ-UHFFFAOYSA-N
hexamethylcyclotrisiloxane,cyclotrisiloxane, hexamethyl,dimethylsiloxane cyclic trimer,unii-epv75l8o0r,ccris 1326,sdk 10,hexamethylcyclohexasiloxane,epv75l8o0r,cyclotrisiloxane, 2,2,4,4,6,6-hexamethyl,dsstox_cid_7185
10914
2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-trioxatrisilinane
C[Si]1(O[Si](O[Si](O1)(C)C)(C)C)C
Applications
Hexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible in strong oxidizing agents.
UN Number UN1325
Beilstein 1766829
Formula Weight 222.47
Percent Purity 97%
Boiling Point 134°C
Melting Point 64°C to 66°C
Quantity 500g
Flash Point 35°C (95°F)
Chemical Name or Material Hexamethylcyclotrisiloxane

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : GZ4705000

TSCA : TSCA