Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethyldisilane, 98+%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
1450-14-2
C6H18Si2
146.38
MFCD00008258
NEXSMEBSBIABKL-UHFFFAOYSA-N
hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane
74057
trimethyl(trimethylsilyl)silane
C[Si](C)(C)[Si](C)(C)C
1450-14-2
C6H18Si2
146.38
MFCD00008258
NEXSMEBSBIABKL-UHFFFAOYSA-N
hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane
74057
trimethyl(trimethylsilyl)silane
C[Si](C)(C)[Si](C)(C)C
Specific Gravity 0.71
Boiling Point 112°C to 114°C
Melting Point 15°C
Flash Point 11°C
Density 0.7100g/mL
Refractive Index 1.4211 to 1.4231
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10mL
Assay Percent Range 98+%
Linear Formula (CH3)3SiSi(CH3)3
Beilstein 04, IV, 4277
Fieser 11,253; 15,164
Formula Weight 146.38
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Hexamethyldisilane
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 215-911-