Alfa Aesar™ Hexamethyldisilazane, Electronic grade, 99+%

Manufacturer: Alfa Aesar™ 4203914

Quantity 25g

 View more versions of this product

Catalog No. AA4203914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 999-97-3
    Linear Formula (CH3)3SiNHSi(CH3)3
    Formula Weight 161.4
    MDL Number MFCD00008259
    UN Number UN3286
    Beilstein 635752
    Merck Index 14,4689
    Synonym HMDS; Bis(trimethylsilyl)amine
    Chemical Name or Material Hexamethyldisilazane
    Grade Electronic Grade

    Specifications

    Formula Weight 161.4
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3286
    RTECS Number JM9230000
    Quantity 25g
    Refractive Index 1.4080
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
    Boiling Point 126°C
    Density 0.774g/mL
    Health Hazard 1 Danger
    Melting Point −78°C
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00008259
    CAS 999-97-3
    Merck Index 14,4689
    Synonym HMDS; Bis(trimethylsilyl)amine
    Flash Point 8° (46°F)
    Sensitivity Moisture Sensitive
    Color Colorless
    Odor Odorless
    Linear Formula (CH3)3SiNHSi(CH3)3
    Beilstein 635752
    Health Hazard 2 GHS H Statement
    H225-H311-H331-H314-H318-H302
    Highly flammable liquid and vapor.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Chemical Name or Material Hexamethyldisilazane
    Assay ≥99%
    Grade Electronic Grade

    GHS H Statement
    H225-H311-H331-H314-H318-H302
    Highly flammable liquid and vapor.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit