Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethyldisiloxane, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
107-46-0
C6H18OSi2
162.38
UQEAIHBTYFGYIE-UHFFFAOYSA-N
trimethyl[(trimethylsilyl)oxy]silane
C[Si](C)(C)O[Si](C)(C)C
107-46-0
C6H18OSi2
162.38
UQEAIHBTYFGYIE-UHFFFAOYSA-N
trimethyl[(trimethylsilyl)oxy]silane
C[Si](C)(C)O[Si](C)(C)C

H2285, 107-46-0, C6H18OSi2

Spectrum™ Hexamethyldisiloxane , also known as Bis(trimethylsilyl) ether or Bis(trimethylsilyl) oxide, is an organosilicon compound used in organic synthesis as a reagent and as a solvent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 25 g
CAS Max % 1
UN Number UN1993