Learn More
Hexamethyleneimine, 98+%, Thermo Scientific Chemicals
Description
Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acids.
Specifications
Melting Point | -37°C |
Density | 0.88 |
Boiling Point | 138°C to 139°C |
Flash Point | 22°C (71°F) |
Odor | Ammonia-like |
Refractive Index | 1.466 |
Quantity | 500 mL |
UN Number | UN2493 |
Beilstein | 1084 |
Solubility Information | Fully miscible in water. |
Show More |
Safety and Handling
Hazard Category | H225-H300-H314-H331 |
Hazard Statement | GHS H Statement H225-H300-H314-H318-H312-H332 Highly flammable liquid and vapour. Fatal if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful in contact with skin. Harmful if inhaled. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXAMETHYLENEIMINE |
EINECSNumber | 203-875-9 |
RTECSNumber | CM3150000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.