Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethyleneimine, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
500mL
Packaging:
Glass bottle
111-49-9
C6H13N
99.17
MFCD00006934
ZSIQJIWKELUFRJ-UHFFFAOYSA-N
hexamethyleneimine,homopiperidine,hexahydroazepine,hexahydro-1h-azepine,perhydroazepine,1h-azepine, hexahydro,hexamethylenimine,azacycloheptane,azepan,hexamethylene imine
8119
CHEBI:32616
azepane
C1CCCNCC1
111-49-9
C6H13N
99.17
MFCD00006934
ZSIQJIWKELUFRJ-UHFFFAOYSA-N
hexamethyleneimine,homopiperidine,hexahydroazepine,hexahydro-1h-azepine,perhydroazepine,1h-azepine, hexahydro,hexamethylenimine,azacycloheptane,azepan,hexamethylene imine
8119
CHEBI:32616
azepane
C1CCCNCC1
Specific Gravity 0.88
Quantity 500mL
Refractive Index 1.4650 to 1.4670
Flash Point 18°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 138.0°C
Melting Point -37.0°C
Assay Percent Range 99%
Beilstein 20, 94
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in strongly alkaline materials, soluble in methanol, ethanol, benzene and ether
Formula Weight 99.17
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Hexamethyleneimine
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger