Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethylenetetramine, 99%, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
Packaging:
Plastic Bottle
Plastic drum
100-97-0
C6H12N4
140.19
VKYKSIONXSXAKP-UHFFFAOYSA-N
methenamine,hexamethylenetetramine,hexamine,urotropine,1,3,5,7-tetraazaadamantane,hexamethylenamine,aminoform,methenamin,urotropin,formamine
4101
CHEBI:6824
C1N2CN3CN1CN(C2)C3
100-97-0
C6H12N4
140.19
VKYKSIONXSXAKP-UHFFFAOYSA-N
methenamine,hexamethylenetetramine,hexamine,urotropine,1,3,5,7-tetraazaadamantane,hexamethylenamine,aminoform,methenamin,urotropin,formamine
4101
CHEBI:6824
C1N2CN3CN1CN(C2)C3
Water 0.5% Max. (K.F.)
Sublimation Point 263.0 °C
Specific Gravity 1.33
Packaging Plastic drum
Flash Point 250°C
Quantity 10kg
Infrared Spectrum Authentic
pH 7 to 10 (1% aq. soln.)
Density 1.3300g/mL
Color White to Yellow
Melting Point 260.0°C
Assay Percent Range 99%
Beilstein 01,583
Fieser 01,427; 02,208; 04,243; 09,234
Merck Index 15,6038
Solubility Information Solubility in water: 895g/L (20°C). Other solubilities: soluble in chloroform: 13.4% (20°C), soluble in ethanol: 2.89% (20°C), soluble in acetone: 0.65% (20°C), insoluble in ether: 0.06% (20°C), soluble in methanol: 7.25% (20°C)
Formula Weight 140.19
Physical Form Powder or Crystals
Percent Purity 99%
Chemical Name or Material Hexamethylenetetramine
Show More Show Less