Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, B&J Brand™, for HPLC, gradient grade, spectrophotometry (not for pesticide residue analysis), >99.9% (>60% n-Hexane), Honeywell Burdick & Jackson

Catalog Number p-7112024
Click to view available options
Quantity:
56 L
200 L
Packaging:
Returnable Steel Drum
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
Absorbance 0.05 AU max. at 225 nm, 0.01 AU max. at 250 nm, 0.005 AU max. at 275 nm, 0.005 AU max. at 300 nm, 0.005 AU max. at 400 nm
CAS Min % >60.0000%
CAS Max % >60.00%, <40.0000%
UN Number UN1208
Beilstein 1730733
Solubility Information 1.4% in Water at 20°C
Formula Weight 86.18
Percent Purity >99.9%
Grade HPLC Gradient
Density 0.659 - 0.673 g/cm3
Boiling Point 68.7°C
Quantity 56 L
Flash Point −26°C
Packaging Returnable Steel Drum
Water 0.01% max.
Chemical Name or Material Hexane, B&J Brand™, for HPLC, gradient grade, spectrophotometry (not for pesticide residue analysis), >99.9% (>60% n-Hexane)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit