Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, HPLC Grade, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
110-54-3
110-54-3
CAS Max % 1
Grade High Purity Solvent
Packaging Amber Glass Bottle
Residue after Evaporation 2 ppm
Thiophene To Pass Test
Water 0.0001
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.