Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes (95% n-hexane), ULTRA RESI-ANALYZED, J.T.Baker™

Click to view available options
Quantity:
1L
4L
110-54-3
C6H14
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
110-54-3
C6H14
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
Assay Percent Range 95% n-hexane
Formula Weight 86.18
Percent Purity 99.5 % min.
Packaging Narrow Mouth Amber Glass Bottle
Quantity 1L
Chemical Name or Material Hexanes (95% n-hexane)