Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, 99% n-hexane, ULTRA RESI-ANALYZED™, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
110-54-3
C6H14
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes
8058
CHEBI:29021
hexane
CCCCCC
110-54-3
C6H14
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes
8058
CHEBI:29021
hexane
CCCCCC

High purity solvents and salts developed for organic residue extraction/concentration procedures

    For use in Gas Chromatography (GC); For Laboratory, Research, or Manufacturing Use
Formula Weight 86.18
Percent Purity 99% n-hexane
Grade ULTRA RESI-ANALYZED™
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.66kg
Flash Point −23.00°C
Quantity 1L
Chemical Name or Material Hexanes