Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, for analysis, mixture of isomers, ACROS Organics™

Manufacturer:  Acros Organics 390740025

 View more versions of this product

Catalog No. AC390740025


Order in Bulk

Request bulk or custom formats

Add to cart

Specifications

Specifications

Hexanes
0.0002 meq/g max.
C6H14
0.01% max. (benzene)
Hex
0.05ppm max.
86.18
Analysis
0.02ppm max.
0.1ppm max.
0.1ppm max.
0.02ppm max.
1.3748 to 1.3810
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Passes Test
0.01% max.
−22°C
69.0°C
-95.0°C
92112-69-1, 110-54-3
0.5ppm max.
CH3(CH2)4CH3
0.1ppm max.
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
0.5ppm max.
Liquid
0.02ppm max.
0.6590g/mL
0.1ppm max.
0.02ppm max.
Glass bottle
0.0005% max.
0.659
0.005% max.
0.1ppm max.
0.31 mPa.s (20°C)
Undesignated
2.5L
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed or concerned: Get medical advice/attention.

Danger

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit