Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
110-54-3
C6H14
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
CHEBI:29021
hexane
CCCCCC
110-54-3
C6H14
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
CHEBI:29021
hexane
CCCCCC

High quality chemicals for laboratory and specialized industrial use

Formula Weight 86.18
Packaging Aluminum Bottle
Specific Gravity 1L = 0.66kg
Flash Point −23.00°C
Quantity 1L
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements
Chemical Name or Material Hexanes