Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
500mL
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

110-54-3
C6H14
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
CHEBI:29021

High quality chemicals for laboratory and specialized industrial use

CAS 110-54-3
Molecular Formula C6H14
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.178
PubChem CID 8058
ChEBI CHEBI:29021
Formula Weight 86.18
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 0.66kg
Flash Point −23.00°C
Quantity 4L
Chemical Name or Material Hexanes
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements
Show More Show Less