Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexanes, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
Specifications
Density | 0.663g/mL at 25°C (typical) |
Boiling Point | 68.7°C |
Quantity | 4 L |
Flash Point | −23.00°C |
Packaging | Aluminum Bottle |
Specific Gravity | 1L = 0.66kg |
Formula Weight | 86.18 |
Grade | Meets ACS reagent chemical requirements |
Chemical Name or Material | Hexanes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction