Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, Reagent, ACS, 98.5%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
110-54-3
C6H14
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
CCCCCC
110-54-3
C6H14
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
CCCCCC

H1010, 110-54-3

Spectrum™ Hexanes, Reagent, ACS , is a mixture of hexane isomers plus methylcyclopentane and is a colorless liquid. It is a used as non-polar solvent since it is low cost, largely unreactive, and relatively safe nature and can be found in specialized glue, as a degreasing agent and to extract cooking oil from seeds. It is used in the laboratory to remove or extract oil and grease contaminant from soil and water sample for analysis. Because it cannot easily be deprotonated it is often used for reactions that involve very strong bases. As an ACS grade Reagent, Hexanes manufactured hexanes is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
CAS Max % 1
UN Number UN1208
Percent Purity 98.5%
Packaging Drum
Residue after Evaporation 1e-005
Sulfur (S) 5e-005
Quantity 200 L