Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes (Technical), Fisher Chemical™

Click to view available options
Quantity:
4L
20L
Packaging:
Aluminum Can
Steel Pail
92112-69-1,110-82-7,64742-49-0
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
Naphtha Solvent
8058
CHEBI:29021
hexane
CCCCCC
92112-69-1,110-82-7,64742-49-0
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
Naphtha Solvent
8058
CHEBI:29021
hexane
CCCCCC

General laboratory use

Additional Information Vapor Pressure: 160mbar at 20°C
Boiling Range 30°C max., 60° to 90°C
Physical Form Liquid
Grade Technical
Density 0.659g/cm³
Identification Pass Test
Packaging Steel Pail
Vapor Pressure 160mbar at 20°C
Flash Point −22°C (71.4°F)
Viscosity 0.31 mPa/s at 20°C
Boiling Point 69°C
Color Colorless
Melting Point -95°C
Quantity 20L
Chemical Name or Material Hexanes
Show More Show Less

DANGER!

Emergency Overview
Extremely flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid