Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes UltimAR, Macron Fine Chemicals™

Click to view available options
Quantity:
1L
4L
Packaging:
Glass
110-54-3
C6H14
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
110-54-3
C6H14
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
Formula Weight 86.18
Packaging Glass
Quantity 1L
Chemical Name or Material Hexanes, (95% n-Hexane)