Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanoic acid, 99%, Acros Organics™

Click to view available options
Quantity:
250g
500g
1kg
2.5kg
Packaging:
Glass bottle
142-62-1
C6H12O2
116.16
MFCD00004421
FUZZWVXGSFPDMH-UHFFFAOYSA-N
caproic acid,capronic acid,n-hexanoic acid,n-caproic acid,butylacetic acid,hexoic acid,pentylformic acid,1-hexanoic acid,n-hexylic acid,n-hexoic acid
8892
CHEBI:30776
hexanoic acid
CCCCCC(=O)O
142-62-1
C6H12O2
116.16
MFCD00004421
FUZZWVXGSFPDMH-UHFFFAOYSA-N
caproic acid,capronic acid,n-hexanoic acid,n-caproic acid,butylacetic acid,hexoic acid,pentylformic acid,1-hexanoic acid,n-hexylic acid,n-hexoic acid
8892
CHEBI:30776
hexanoic acid
CCCCCC(=O)O
Viscosity 3.23 mPa.s (20°C)
Acidity 473 to 493mg KOH/g
Specific Gravity 0.92
Flash Point 104°C
Quantity 2.5kg
Packaging Glass bottle
Melting Point -3.0°C
Refractive Index 1.4150 to 1.4180
Infrared Spectrum Authentic
Color Colorless to Yellow
Boiling Point 202.0°C to 203.0°C
Density 0.9200g/mL
Assay Percent Range 99%
Linear Formula CH3(CH2)4CO2H
Beilstein 02, 321
Merck Index 15, 1761
Solubility Information Solubility in water: 1.1% (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 116.16
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Hexanoic acid
Show More Show Less

GHS P Statement Causes severe skin burns and eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger