Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanoic anhydride, 97%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
2051-49-2
C12H22O3
214.3
MFCD00009509
PKHMTIRCAFTBDS-UHFFFAOYSA-N
hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride
74918
hexanoyl hexanoate
CCCCCC(=O)OC(=O)CCCCC
2051-49-2
C12H22O3
214.3
MFCD00009509
PKHMTIRCAFTBDS-UHFFFAOYSA-N
hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride
74918
hexanoyl hexanoate
CCCCCC(=O)OC(=O)CCCCC
Specific Gravity 0.926
Boiling Point 246°C to 248°C
Flash Point 130°C
Melting Point -40°C
Refractive Index 1.4270 to 1.429
Color Colorless to Yellow
Infrared Spectrum Authentic
Density 0.9260g/mL
Quantity 250g
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula [CH3(CH2)4CO]2O
Beilstein 02, 324
Solubility Information Solubility in water: 72g/L. Other solubilities: soluble in ethanol and ether
Formula Weight 214.3
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Hexanoic anhydride, 99%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 218-121-4

TSCA : TSCA