Alfa Aesar™ Hexanoic anhydride, 98%

Manufacturer: Alfa Aesar™ B2375630

Quantity 250g

 View more versions of this product

Catalog No. AAB2375630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2051-49-2
    Molecular Formula C12H22O3
    Linear Formula [CH3(CH2)4CO]2O
    Formula Weight 214.31
    MDL Number MFCD00009509
    UN Number UN3265
    Beilstein 1776561
    Synonym Caproic anhydride
    Chemical Name or Material Hexanoic anhydride

    Specifications

    Formula Weight 214.31
    TSCA TSCA
    Linear Formula [CH3(CH2)4CO]2O
    MDL Number MFCD00009509
    CAS 2051-49-2
    UN Number UN3265
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Odor Unpleasant
    Molecular Formula C12H22O3
    Beilstein 1776561
    Synonym Caproic anhydride
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 128°C (262°F)
    Boiling Point 246° to 248°C
    Melting Point −40°C
    Quantity 250g
    Chemical Name or Material Hexanoic anhydride
    Assay 98%

    Description

    Applications
    It is used as a reactant in the synthesis of acremomannolipin A. It is used in green synthesis of esters of acyclovir (acyclovir prodrugs), in the preparation of hexanoyl-modified chitosan nanoparticles, chitosan-based polymeric surfactants via N-acylation of chitosans.

    Solubility
    Hydrolyzes in water.

    Notes
    Moisture sensitive. Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, bases, water/ moisture.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit