Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanophenone 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
942-92-7
C12H16O
176.26
MFCD00009512
MAHPVQDVMLWUAG-UHFFFAOYSA-N
hexanophenone,caprophenone,1-hexanone, 1-phenyl,1-phenyl-1-hexanone,n-hexanophenone,amyl phenyl ketone,hexaphenone,pentyl phenyl ketone,4-ethylbutyrophenone,n-amylphenyl ketone
70337
1-phenylhexan-1-one
CCCCCC(=O)C1=CC=CC=C1
942-92-7
C12H16O
176.26
MFCD00009512
MAHPVQDVMLWUAG-UHFFFAOYSA-N
hexanophenone,caprophenone,1-hexanone, 1-phenyl,1-phenyl-1-hexanone,n-hexanophenone,amyl phenyl ketone,hexaphenone,pentyl phenyl ketone,4-ethylbutyrophenone,n-amylphenyl ketone
70337
1-phenylhexan-1-one
CCCCCC(=O)C1=CC=CC=C1
Melting Point 25.0°C to 26.0°C
Refractive Index 1.5105
Quantity 25g
Packaging Glass bottle
Specific Gravity 0.95
Infrared Spectrum Authentic
Flash Point >112°C
Color Yellow
Density 0.9500g/mL
Boiling Point 265.1°C
Assay Percent Range 98%
Linear Formula C6H5CO(CH2)4CH3
Beilstein 07,II,257
Formula Weight 176.26
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material Hexanophenone, 98%
Show More Show Less

EINECSNumber : 213-394-6

TSCA : TSCA