Learn More
Hexanoyl-Lysine adduct Mouse anti-Non-species specific, Alexa Fluor 488, Clone: 5D9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP259362AF488
Catalog No. NB259362488
Add to cart
Description
Description
Hexanoyl-Lysine adduct Monoclonal specifically detects Hexanoyl-Lysine adduct in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Hexanoyl-Lysine adduct
5D9
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Purified
Synthetic Hexanoyl modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Nonspecies specific
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1
100 ul
Primary
Specific for Hexanoyl-Lysine adduct (HEL) modified peptides and proteins. Does not detect free Hexanoyl-Lysine. Does not cross-react with Acrolein, Crotonaldehyde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.