Learn More
Hexanoyl-Lysine adduct Mouse anti-Non-species specific, Biotin, Clone: 500000000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259363B
Catalog No. NBP259363B
Add to cart
Description
Description
Hexanoyl-Lysine adduct Monoclonal antibody specifically detects Hexanoyl-Lysine adduct in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Hexanoyl-Lysine adduct
500000000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
Synthetic Hexanoyl modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
Mouse
IgG1
100 ul
Primary
Specific for Hexanoyl-Lysine adduct (HEL) modified peptides and proteins. Does not detect free Hexanoyl-Lysine. Does not cross-react with Acrolein, Crotonaldehyde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.