Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexanoyl-Lysine adduct Mouse anti-Non-species specific, Clone: 5.00E+08, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259363
Description
Hexanoyl-Lysine adduct Monoclonal specifically detects Hexanoyl-Lysine adduct in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Hexanoyl-Lysine adduct | |
Monoclonal | |
Unconjugated | |
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
Synthetic Hexanoyl modified Keyhole Limpet Hemocyanin (KLH). | |
100 μg | |
Primary | |
Nonspecies specific | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
500000000 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Mouse | |
Protein G purified | |
RUO | |
Specific for Hexanoyl-Lysine adduct (HEL) modified peptides and proteins. Does not detect free Hexanoyl-Lysine. Does not cross-react with Acrolein, Crotonaldehyde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins. | |
Store at -20C. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only