Learn More
Hexanoyl-Lysine adduct Mouse, ATTO 565, Clone: 5D9, Abnova
Mouse Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB19259
Description
Hexanoyl-Lysine adduct Monoclonal antibody specifically detects Hexanoyl-Lysine adduct in samples. It is validated for WB, ELISA, ICC, IF, Flow Cyt.Specifications
Hexanoyl-Lysine adduct | |
5D9 | |
Liquid | |
Synthetic Hexanoyl modified Keyhole Limpet Hemocyanin (KLH) | |
100 μg | |
Primary |
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
ATTO 565 | |
Mouse | |
IgG1 | |
Store at -20°C | |
Monoclonal |