Alfa Aesar™ Hexaphenylditin, 97+%

Manufacturer: Alfa Aesar™ 7114306

Quantity 5g

 View more versions of this product

Catalog No. AA7114306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1064-10-4
    Molecular Formula (C6H5)6Sn2
    Formula Weight 700.02
    MDL Number MFCD00014070
    UN Number UN3146
    Chemical Name or Material Hexaphenylditin

    Specifications

    Formula Weight 700.02
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Boiling Point 280°C (dec.)
    Physical Form Powder
    MDL Number MFCD00014070
    Quantity 5g
    UN Number UN3146
    Odor Characteristic
    Melting Point 232.5°C
    Molecular Formula (C6H5)6Sn2
    CAS 1064-10-4
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
    Chemical Name or Material Hexaphenylditin
    Assay ≥97%

    GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit