Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hexarhodium hexadecacarbonyl, 98%

Click to view available options
Quantity:
0.1g
0.25g
0.5g
28407-51-4
C16O16Rh6
1065.593
MFCD00011206
SZQABOJVTZVBHE-UHFFFAOYSA-N
hexarhodium hexadecacarbonyl,hexarhodiumhexadecacarbonyl,rhodium 0 carbonyl,rh6 co 16,hexarhodium 0 hexadecacarbonyl,hexadecacarbonylhexarhodium,hexarhodium 0 hexadecacarbonyl, rh 57-60 % approx.
10866043
carbon monoxide;rhodium
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Rh].[Rh].[Rh].[Rh].[Rh].[Rh]
28407-51-4
C16O16Rh6
1065.593
MFCD00011206
SZQABOJVTZVBHE-UHFFFAOYSA-N
hexarhodium hexadecacarbonyl,hexarhodiumhexadecacarbonyl,rhodium 0 carbonyl,rh6 co 16,hexarhodium 0 hexadecacarbonyl,hexadecacarbonylhexarhodium,hexarhodium 0 hexadecacarbonyl, rh 57-60 % approx.
10866043
carbon monoxide;rhodium
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Rh].[Rh].[Rh].[Rh].[Rh].[Rh]
Melting Point >300°C
Quantity 0.1g
UN Number UN3466
Formula Weight 1065.6
Percent Purity 98%
Chemical Name or Material Hexarhodium hexadecacarbonyl

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.

TSCA : TSCA