HexArmor™ NXT™ Series Cut-Resistant Gloves

Manufacturer: Performance Fabrics, IncSupplier Diversity Partner 10302M

 View more versions of this product

Catalog No. 19-163-509

  Check Availability
Add to Cart

    Specifications

    Cut Resistance Rating Level 5
    Glove Cuff Style Knit
    Hand Ambidextrous
    For Use With (Application) food service, cutting and processing, deboning applications, knife/blade handling, Plastics trimming
    Material Super Fabric™, Coretek™ Fiber
    Size Medium
    Coating Material Nitrile
    Color White, gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit