Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HexArmor™ NXT™ Series Cut-Resistant Gloves

$52.50

Specifications

Coating Material Nitrile
Color Gray, White
Cut Resistance Rating Level 5
Material Coretek™ Fiber, SuperFabric™
For Use With (Application) food service, cutting and processing, deboning applications, knife/blade handling, Plastics trimming
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-163-506
View Documents
HexArmor™
10302XXS
2X-Small
Each for $52.50
Add to cart
 
19-163-511
View Documents
HexArmor™
10302XL
X-Large
Each for $52.50
Add to cart
 
19-163-508
View Documents
HexArmor™
10302S
Small
Each for $52.50
Add to cart
 
19-163-509
View Documents
HexArmor™
10302M
Medium
Each for $52.50
Add to cart
 
19-163-512
View Documents
HexArmor™
10302XXL
2X-Large
Each for $52.50
Add to cart
 
Specifications

Specifications

Nitrile
Level 5
food service, cutting and processing, deboning applications, knife/blade handling, Plastics trimming
Ambidextrous
Gray, White
Coretek™ Fiber, SuperFabric™
Knit
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit