HexArmor™ Protective Aprons

Exceptional protection against blunted poultry knives

Manufacturer: Performance Fabrics, IncSupplier Diversity Partner AP322

 View more versions of this product

Catalog No. 19-160-617

  Check Availability
Add to Cart

    Specifications

    Color Gray, black
    Material SuperFabric™, Resin Coated Nylon
    Width (Metric) 61cm
    Width (English) 24 in.
    Includes Adjustable straps
    Certifications/Compliance ISEA CE Level 5
    For Use With (Application) Veneer pulling, food processing, filing/grinding
    Size Universal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit