Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HexArmor™ Protective Aprons
SDP

Catalog Number 191303755
Click to view available options
Dimensions (L x W):
22 x 22 in.
29 x 29 in.
30 x 24 in.
Size:
Universal
Thickness:
Double layer
Single layer
This item is not returnable. View return policy
This item is not returnable. View return policy

Exceptional protection against blunted poultry knives

Cut, puncture, and abrasion resistant aprons protect body from accidents on the job site. Help workers to stay warm, safe, and comfortable with less fatigue.

  • Offer ISEA Level 5 cut resistance
  • No metal disks or links to repair
  • Lightweight—only 1/10 the weight of traditional chain mail safety wear
  • Safe for machine wash and sanitizers
  • Available in two lengths; both include adjustable strap

1These aprons are cut and/or puncture resistant, not cut or puncture proof. Always conduct a safe test with your application to determine if the product is suited for your needs.

Size Universal
Material SuperFabric™
Color Black
Thickness Double layer
Includes Adjustable straps
For Use With (Application) Ideal for meat processing and packing, grinding and cutting/puncture applications
Certifications/Compliance ISEA CE Level 5
Dimensions (L x W) 22 x 22 in.
Width (English) 20 in.
Width (Metric) 50 cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit