Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEXB Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen HEXB
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096185
View Documents
Novus Biologicals
NBP304920100UL
100 μg Each for $320.71
Add to cart
 
NB096186
View Documents
Novus Biologicals
NBP30492020UL
20 μg Each for $138.37
Add to cart
 
Description

Description

HEXB Polyclonal antibody specifically detects HEXB in Mouse samples. It is validated for Immunofluorescence
Specifications

Specifications

HEXB
Unconjugated
Rabbit
Neurodegeneration
PBS with 50% glycerol, pH7.3.
3074
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence
Purified
RUO
Mouse
beta-hexosaminidase subunit beta, Beta-N-acetylhexosaminidase subunit beta, Cervical cancer proto-oncogene 7 protein, EC 3.2.1.52, ENC-1AS, HCC-7, hexosaminidase B (beta polypeptide), Hexosaminidase subunit B, N-acetyl-beta-glucosaminidase subunit beta
Recombinant fusion protein containing a sequence corresponding to amino acids 277-556 of human HEXB (NP_000512.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit