Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexestrol, ≥98%, MilliporeSigma™ Supelco™

Hexestrol is a synthetic nonsteroidal estrogen, which can be used in human medicine as well as a growth promoter in cattle.

$59.60 - $201.50

Specifications

CAS 84-16-2
Percent Purity ≥98%
Grade Analytical Standard
Linear Formula C18H22O2
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-5662
View Documents
MilliporeSigma Supelco
H77531G
1 g
Each for $59.60
Add to cart
 
11-101-5663
View Documents
MilliporeSigma Supelco
H77535G
5 g
Each for $201.50
Add to cart
 
Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

84-16-2
Analytical Standard
Neat
≥98%
C18H22O2
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit