Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexokinase 2 Mouse anti-Human, Canine, Allophycocyanin/Cy7, Clone: OTI4C5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270891APCCY7

Catalog No. N270891APC7


Add to cart

Description

Description

Hexokinase 2 Monoclonal antibody specifically detects Hexokinase 2 in Human, Canine samples. It is validated for Flow Cytometry.
Specifications

Specifications

Hexokinase 2
OTI4C5
Flow Cytometry
Affinity Purified, Conjugated
HK2
Full length recombinant protein of human Hexokinase 2 (NP_000180) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication
Primary
3099.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit