Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexokinase 2 Mouse anti-Human, DyLight 350, Clone: 1A7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104296UV
Description
Hexokinase 2 Monoclonal specifically detects Hexokinase 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Hexokinase 2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HK2 | |
Recombinant human Hexokinase 2 (1-917aa) purified from E. coli | |
0.1 mL | |
Cell Cycle and Replication | |
3099 | |
Human | |
Purified |
Western Blot, ELISA | |
1A7 | |
Western Blot, ELISA | |
DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human Hexokinase 2 mAb, clone 1A7, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Hexokinase 2 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title