Learn More
BioVision IncSupplier Diversity Partner HEXOKINASE ACTIVITY KIT-FLUORO DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K769100
PicoProbe Hexokinase Activity Assay Kit (Fluorometric):Hexokinase (EC 1.1.1.49; also called HK, 6-Phosphate Glucose Kinase, ATP: D-Hexose 6-Phosphotransferase, ATP-dependent Hexokinase) is found in many organisms including bacteria, plants and mammals. Hexokinases play an important role in glucose metabolism. Hexokinase deficiency leads to diseases such as X-linked muscular dystrophy and rare autosomal recessive hemolytic anemia. On the other hand, increased hexokinase activity is detected in various human tumors and is associated with metastasis. BioVisions PicoProbe Hexokinase Activity Assay kit provides a quick and easy method for monitoring HK activity in a wide variety of samples. In this assay, HK converts glucose into glucose-6-Phosphate, which in turn undergoes a series of reactions and reduce PicoProbe to generate intense fluorescent product (Ex/Em = 535/587 nm). The assay is simple, sensitive and high-throughput adaptable and can detect as low as 2 ��U of HK activity.
Catalog No. 50-111-9892