Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexokinase1 Mouse anti-Human, Janelia Fluor 646, Clone: 4D7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP104295JF646

Catalog No. NBP14295646


Add to cart

Description

Description

Hexokinase1 Monoclonal antibody specifically detects Hexokinase1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Hexokinase1
4D7
Immunocytochemistry/Immunofluorescence
Purified
HK1
Recombinant human Hexokinase1 (1-917aa) purified from E. coli
Protein G purified
Cancer
Primary
3098
Human
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Brain form hexokinase, EC 2.7.1, EC 2.7.1.1, glycolytic enzyme, hexokinase 1, Hexokinase type I, hexokinase-1, HK I, HK1-ta, HK1-tb, HK1-tc, HKI, HXK1
Mouse
IgG2a Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
The antibody recognizes all four isoforms of Hexokinase (1~4) in recombinant protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit