Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexosaminidase A/HEXA Mouse anti-Human, Alexa Fluor™ 594, Clone: 20F1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242629AF594

Catalog No. NP242629A94


Add to cart

Description

Description

Hexosaminidase A/HEXA Monoclonal antibody specifically detects Hexosaminidase A/HEXA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

Hexosaminidase A/HEXA
20F1
Western Blot, Flow Cytometry, ELISA
Conjugated, Purified
HEXA
Recombinant human HEXA (89-529aa) purified from E. coli
Protein A purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
beta-hexosaminidase subunit alpha, Beta-N-acetylhexosaminidase subunit alpha, EC 3.2.1, EC 3.2.1.52, hexosaminidase A (alpha polypeptide), Hexosaminidase subunit A, MGC99608, N-acetyl-beta-glucosaminidase subunit alpha, TSD
Mouse
IgG2a Lambda
100 μL
Primary
3073.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit